VWR® Critical Swab® Swab, Medium Foam Head, Wooden Shaft
Supplier: VWR
VWR® Critical Swab®
1806-WF VWR 1806-WFVWR
CA89031-274CS 1898.09 CAD
CA89031-274 89031-274
VWR® Critical Swab® Swab, Medium Foam Head, Wooden Shaft
Swabs Swabs for Sampling
Disposable swab features 100% polyurethane foam with 100ppi and a wood shaft.
  • Polyurethane foam head
  • Wood shaft
  • Disposable