SGE BP1 GC Capillary Columns
Supplier: Trajan Scientific and Medical
054022 054045 054056 054068 054069 054070 054073 054085 054092 054806 054807 054808 054809 054810 054812 054813 054815 054818 054820 054096 054095 054090 054087 054086 054082 054072 054071 054066 054065 054064 054062 054061 054059 054054 054053 054051 054050 054048 054047 054046 054044 054043 054041 054040 054039
14225-864EA 841.95 CAD
14225-864 14225-868 14225-870 14225-872 14225-874 14225-876 14225-878 14225-884 14225-888 14226-056 14226-058 14226-060 14226-062 14226-064 14226-068 14226-070 14226-072 14226-074 14226-078 60362-094 60362-096 60362-100 60362-104 60362-106 60362-108 60362-114 60362-116 60362-120 60362-122 60362-124 60362-126 60362-128 60362-132 60362-136 60362-138 60362-142 60362-144 60362-148 60362-150 60362-152 60362-154 60362-156 60362-160 60362-162 76409-322
SGE BP1 GC Capillary Columns
The SGE BP1 GC Capillary Column provides a great non-polar option for GC analyses. Recommended for: Hydrocarbons, aromatics, pesticides, phenol, herbicides, amines.
  • Classic dimethyl polysiloxane technology with high temperature cross-linking
  • Excellent general-purpose GC column
  • Low bleed
  • Non-polar
  • Suitable for all routine analyses
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.