SAFECAP® Capillary Tubes, Self-Sealing Mylar® Wrapped
Supplier: Drummond
H75-2000M P75-2000M R60-2000M
77497-226BX 657.28 CAD
77497-226 77497-228 77497-222
SAFECAP® Capillary Tubes, Self-Sealing Mylar® Wrapped
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
SAFECAP® is a single-use, Triple Mylar® wrapped capillary tube with a super absorbent plug that automatically seals on contact with blood.
  • Unique
  • Self sealing capillary tubes
  • Mylar® wrap minimizes risk of exposure to HIV, hepatitis, and other bloodborne pathogens
  • Case of 20 vials contains 100 tubes each
The Mylar® wrap significantly reduces exposure to potentially life-threatening blood-borne pathogens and glass fragments, while the self-sealing plug eliminates manually sealing with clay and reduces centrifuge blowouts.