PurSwab® Standard Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1804-PF 1804-PFB 1804-PF 1804-PFB
CA10806-020CS 963.75 CAD
CA10806-020 CA10806-021 10806-020 10806-021
PurSwab® Standard Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Lint-free, nonabrasive, shred-resistant swabs are ideal for precision cleaning
Made in the USA.
The 100ppi polyurethane foam tip is securely bonded to the polypropylene shaft.
Certifications: CE marked.