Contamination Control Swab, Foam Head, Flexible Tip, Foamtec International WCC
Supplier: Foamtec
HT1001
10810-862CS 3961.12 CAD
10810-862
Contamination Control Swab, Foam Head, Flexible Tip, Foamtec International WCC
Swabs Swabs for Cleaning
Designed to meet demanding cleanroom applications
Can be used with alcohol, Freon® 113 fluorocarbon, acetone, and other solvents for assembly cleaning, and for epoxy application. Flexible-tip swab features a 100ppi polyurethane head and a polypropylene handle.
Ordering information: Packaged in polybags.