Rtx®-CLPesticides and Rtx-CLPesticides2 GC Columns, Restek
Supplier: Restek
11139 11140 11324 11340 11141 11325 11123 11323 11123-600 11139-600 42301 42302 11321 11326 42102 11120 11136 11126 11324-600
RK11139EA 1920.4 CAD
RK11139 RK11140 RK11324 RK11340 10057-830 10057-832 10803-178 10803-180 10849-656 10849-658 10849-894 10849-896 10850-008 10850-010 10850-070 10850-388 10850-390 10850-658 10851-988
Rtx®-CLPesticides and Rtx-CLPesticides2 GC Columns, Restek
Chromatography Columns
Improved resolution and faster analyses make these the ideal pesticide columns.
  • Application-specific columns
  • Designed to overcome coelutions and analyte breakdown typically encountered in chlorinated pesticide analyses
  • Improved resolution
  • Faster analytical results
The application-specific columns for organochlorine pesticides and herbicides exceed performance criteria for US EPA methods 8081, 608, and CLP. Columns are specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide analyses. In order to provide reliable identification without GC/MS, more accurate qualitative data can be obtained by achieving baseline resolution of the 20 target analytes. Column bleed is extremely low at temperatures greater than 300 °C (572 °F), which is critical for the removal of high-boiling compounds commonly encountered in pesticide/PCB extracts. Analysis time is typically less than 25 minutes.