Primate MD's™ Anti-Inflammatory
Supplier: Bio-Serv
F07096 MD650-90 F07096 MD650-90
76285-256PK 170.64 CAD
76285-256 76419-512 CA76285-256 CA76419-512
Primate MD's™ Anti-Inflammatory
Animal Supplies Animal Feed
Chewable, flavored, medicated tablet replaces more stressful dosing methods.
  • Tablet consistency ensures accurate dosing
  • Scored 5 gm tablet for flexible dose administration
  • Cold tableting process eliminates drug degradation
  • No heat or water used in processing
  • Highly palatable sugar-based formula with excellent drug masking capabilities
  • Reduces time, labor and stress compared to traditional dosing methods
  • MD's can be customized to meet specific requirements
  • Shelf life: 6 to 24 months as dictated by the drug expiration date
Base Meal: Sucrose, dextrose, mineral oil, artificial and natural flavors, calcium silicate, magnesium stearate.
.