VWR® Talon® Heavy-Duty Support Stands
Supplier: VWR International
97035-510 97035-512 97035-514 97035-516 97035-518 97035-520
97035-510EA 795.86 CAD
97035-510 97035-512 97035-514 97035-516 97035-518 97035-520
VWR® Talon® Heavy-Duty Support Stands
Retort Stands
Heavy-duty support stands permit cylindrical vessels up to 30.5 cm (12") in diameter to sit close to the support rod.
  • Cast iron base with black enamel finish
  • Optional stainless steel support rods
  • Rubber cushions under each support stand corner
Ideal for mounting stirring motors for heavy equipment
Accessories include extra stainless steel support rods and two choices of clamps. The support stand clamp (58950-660) is designed with a precision fit to accept a 13mm (1/2") diameter support shaft and a 16mm (5/8") diameter support rod on a support stand.More Product Information