Rtx®-Wax GC Columns, Restek
Supplier: Restek
12421 12458 12423 12424 12426 12427 12435 12436 12438 12439 12440 12441 12442 12452 12454 12457 12408 12420 12451 12455 12455 41602 41603
89048-602EA 800.37 CAD
89048-602 89048-644 89048-606 89048-608 89048-612 89048-614 89048-616 89048-618 89048-622 89048-624 89048-626 89048-628 89048-630 89048-636 89048-638 89048-642 89048-596 89048-600 89048-634 89048-640 AP400000-002 10059-538 10059-540 10059-542
Rtx®-Wax GC Columns, Restek
Chromatography Columns
Highly efficient and inert general-purpose PEG columns are ideal for FAMEs, solvents, BTEX aromatics, and flavor compounds.
  • Excellent selectivity for compounds of intermediate to high polarity
  • Equivalent to USP G14, G15, G16, G20, and G39 phases
  • Temperature range: 20 to 250 °C (68 to 482 °F)
Columns feature an extended operating temperature range to allow for the analysis of compounds having a wide volatility range, and to ensure low bleed at temperatures as high as 250 °C (482 °F).