Rxi®-5ms Capillary Columns, Restek
Supplier: Restek
13409 13441 13442 13481 13454 13402 13420 13422 13423 13426 13438 13439 13440 13456 13457 13467 13470 13411 13421 13424 13425 13427 13435 13436 13437 13450 13451 13452 13453 13455 13497 13498 13499 13423-600 13401 13455-600
89048-014EA 1054.2 CAD
89048-014 89048-046 89048-048 89048-070 RK13454 89048-012 89048-018 89048-022 89048-024 89048-030 89048-040 89048-042 89048-044 89048-062 89048-064 89048-066 89048-068 89048-016 89048-020 89048-026 89048-028 89048-032 89048-034 89048-036 89048-038 89048-050 89048-052 89048-054 89048-056 89048-060 89048-072 89048-074 89048-076 10058-214 10850-458 10852-042
Rxi®-5ms Capillary Columns, Restek
Chromatography Columns
Inert, low-bleed columns for reliable column-to-column results from low-level GC/MS analyses provide reproducible retention and selectivity.
  • Low bleed
  • Reliable column-to-column results
  • General-purpose columns
  • Inert properties
Designed for semi-volatiles, drugs, pesticides, PCB congeners, solvent impurities, phenols, amines, and other general-purpose analyses. Equivalent to USP G27 Phase (Crossbond® 5% diphenyl, 95% dimethyl polysiloxane). Temperature range: −60 to 330/350 °C (−76 to 626/662 °F).