Absorbent Liners, Jaece
Supplier: Jaece
AL2050/6 AL20300 AL20600 AL20501
82002-604CS 503.35 CAD
82002-604 82002-606 82002-608 10159-110
Absorbent Liners, Jaece
Surface Protectors Bench Protectors
Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals.
  • Cushions surfaces while absorbing liquids
  • Can be cut to size
Keeps spills from spreading and protects benchtops, floors, and carts.
Ordering information: Liner 82002-606 is packaged one roll per case.