Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D9860005 D9860012 D1069324 D1069329 D9860008 D9860011 D9860002 D9860003
54112-085EA 908.42 CAD
54112-085 54112-090 54112-100 54112-256 66110-950 66110-954 66110-956 66110-958
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Surface Protectors Bench Protectors
Put the benefits of Teflon® resin on any flat surface with this pressure-sensitive overlay
Available with two different FEP bonded support sheets. Aluminum-backed overlay can withstand temperatures up to 205°C (400°F). Vinyl-backed overlay can withstand temperatures up to 93°C (200°F).
Create a non-stick, easy-to-clean, easy-to-apply work surface. Non-contaminating and chemically inert. Ideal as bench-top covering or release liner for inside surfaces. Overlay is made of 0.02mm (0.001") thick Teflon® resin. Easily cut with scissors.