DURA-STAT® Soft Work Surfaces, Duracote
Supplier: Duracote
TM8B0203 TM8B0204 TM8B0440 TM8T0203 TM8B0240
56360-120CS 158.82 CAD
56360-120 56360-122 56360-124 56360-126 56360-130
DURA-STAT® Soft Work Surfaces, Duracote
Surface Protectors Bench Protectors
These soft, static-dissipative work surfaces have a resistance-to-ground measure of 107 to 108 ohms at 10V, 12% relative humidity
In accordance with the ESD Association, they are tested to EOS/ESD S4.1 to meet industry guidelines for proper electrostatic discharge. Work surfaces are engineered to meet the requirements of MIL-W-87893. The unique construction provides the same resistance-to-ground measurement from any point on the material. Lightweight and flexible, these multi-layer vinyl surfaces are available in light blue or beige. The top surface is semi-conductive, non-tacky, and dirt-More Product Information