SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Supplier: Thermo Scientific
6A24ST26 6A20ST20
TS6A24ST26PK 209.37 CAD
TS6A24ST26 TS6A20ST20
SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Septa
Level 2 septa are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
  • Autoclavable
  • Single use
  • Non sterile
Choose all-purpose silicone/PTFE septa manufactured with a shore value of 50 to reduce autosampler needle issues.
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables.More Product Information