CleanTips® Polyester Alpha® Mini Swab, Texwipe®
Supplier: Texwipe
CleanTips® Alpha®
TX754B
TWTX754BCS 953.86 CAD
TWTX754B
CleanTips® Polyester Alpha® Mini Swab, Texwipe®
Swabs Swabs for Cleaning
The Alpha® Mini Swab is an excellent swab for cleaning small areas
The compact polypropylene handle provides support and control. Low particle generation and extractable levels. Excellent chemical resistance.
It's cleanroom laundered, double-layered, knitted Alpha® polyester head is extremely clean. Swabs feature 100% continuous-filament polyester substrate that is heat-molded (thermally-bonded) onto the handle, sealing exposed edges for reduced fiber release and eliminating the need for adhesives. Mini tip with a rigid paddle
Ordering information: Available in bag or case quantities.