Puritan® Small Cotton Tipped Applicators, Wooden Handle, Puritan Medical Products
Supplier: Puritan Medical Products
896-WC 876-WC 896-WC 876-WC
CA10805-186CS 2258.04 CAD
CA10805-186 CA82030-754 CA89133-812 82030-754 10805-186 89133-812
Puritan® Small Cotton Tipped Applicators, Wooden Handle, Puritan Medical Products
Swabs Swabs for Sampling
For cleaning in sensitive environments.
Lint-free and absorbent single-end cotton tip. Wood shaft.
Certifications: CE marked.
Ordering information: Packaged 100 swabs per bag, 10 bags per box, 10 boxes per case.