Formalin, VWRClick, neutral buffered prefill
Supplier: VWR International
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
77507-050
77507-052
77507-054
77507-056
77507-058
77507-042
77507-044
77507-046
77507-048
77507-040
77507-060
77507-050CS
291.72
CAD
77507-050
77507-052
77507-054
77507-056
77507-058
77507-042
77507-044
77507-046
77507-048
77507-040
77507-060
Formalin, VWRClick, neutral buffered prefill
Formalin
VWR®, part of Avantor has industry leading Anatomical Pathology prefills available for your lab.
- VWRClick containers produce an audible “click” sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.
- Packaged in HistoPack carrier for easy distribution or in bulk.
- Containers are 95kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.
- "Click" available in 20 ml, 40 ml, 60 ml, 90 ml, 120 ml sizes.
Certifications: ISO 13485 Certified
Formula:
CH₂O Boiling Pt: 100 °C (212 °F) Melting Pt: –92 °C (–133 °F) Flash Pt: 93.3 °C (199.9 °F) |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...