VWR® Critical Swab® Smooth Cotton Double Tipped Swab, Paper Handle
Supplier: VWR International
CA89031-290 89031-290
CA89031-290CS 585.75 CAD
CA89031-290 89031-290
VWR® Critical Swab® Smooth Cotton Double Tipped Swab, Paper Handle
Swabs Swabs for Sampling
Swab features a paper stem and two tips composed of lintless, highly absorbent cotton.
  • Antistatic packing
  • Lintless, highly absorbent cotton
Packaged in an antistatic leaf pack.
Certifications: CE marked.
Ordering information: Packaged as 50 leaves of 25 swabs per leave.