Puritan® Cap-Shure® Sterile Swab with Cap, Puritan Medical Products
Supplier: Puritan Medical Products
25-8071PCEC 25-8071PCEC 25-8061WCEC
CA82029-732CS 2292.75 CAD
CA82029-732 82029-732 82029-070
Puritan® Cap-Shure® Sterile Swab with Cap, Puritan Medical Products
Swabs Transport Swabs
Cotton-tipped sterile swabs are designed for convenient and reliable specimen, DNA, or evidence collection
Clear cap is aerated for air-drying and transporting samples. This swab is available with wood or plastic shaft. Individually wrapped.