Rtx®-1301 GC Columns, Restek
Supplier: Restek
16023 16052 16053 16069 16072 16075 16021 16024 16040 16056 16058 16070 16020 16026 16038 16039 16054 16057 16085 16085-126 16088 16091 16055 16066 16076 16025 16016 16092 16085-600 16043 16093 16085-129
RK16023EA 1165.83 CAD
RK16023 RK16052 RK16053 RK16069 RK16072 RK16075 RK16021 RK16024 RK16040 RK16056 RK16058 RK16070 RK16020 RK16026 RK16038 RK16039 RK16054 RK16057 RK16085 RK16085-126 RK16088 RK16091 RK16055 RK16066 RK16076 RK16025 10803-098 10803-100 10848-758 10849-276 10849-358 10849-418 10850-226 10850-228 10850-502 75850-010
Rtx®-1301 GC Columns, Restek
Chromatography Columns
General-purpose columns designed for use with residual solvents, alcohols, oxygenates, and volatile organic compounds.
  • Low bleed
  • Longer column lifetime
  • Increased inertness
  • General-purpose columns
The cyanosilicone-bonded stationary phase offers low bleeds, long lifetimes, and increased inertness.
Phase composition: 6% cyanopropylphenyl/94% dimethyl polysiloxane.
Polymer is fully characterized to ensure long-term reproducibility, column-to-column consistency, and low bleed even with sensitive detectors such as ECDs and MSDs. Equivalent to USP G43 phase.