Rtx®-50 Phase Columns, Restek
Supplier: Restek
10538 10542 10553 10557 10567 40502 10524 10526 10552 10554 10522 10523 10535 10536 10539 10543 10558 10569 10570 10540 10555 10551 40511
RK10520EA 701.7 CAD
RK10520 RK10538 RK10542 RK10553 RK10557 RK10567 RK40502 RK10524 RK10526 RK10537 RK10552 RK10554 RK10521 RK10522 RK10523 RK10535 RK10536 RK10539 RK10543 RK10558 RK10569 RK10570 RK10540 RK10550 RK10555 RK10551 10850-704 10850-710
Rtx®-50 Phase Columns, Restek
Chromatography Columns
For analysis of pesticides, herbicides, phthalate esters, cholesterols, triglycerides, and rosin acids.
  • Low bleed
  • Longer column lifetime
  • Polymer is fully characterized to ensure long-term reproducibility
Can be used as a confirmational column for many environmental applications including EPA methods 604 (phenols) and 8060 (phthalate esters) and for the analysis of EPA method 608 (pesticides).
Fused silica columns are coated with crossbonded 50% methyl/50% phenyl polysiloxane stationary phase. The high thermal stability of these columns allows dual column analysis with common phases such as Rtx®-1 and Rtx®-5 for simultaneous run conditions and high column bake-out temperatures to remove contamination. Restek's Crossbond® procedure produces columns with lower bleed, longer lifetime, and column rejuvenation through solvent washing. No residual catalyst remains in the polymer which could cause degradation and increase bleed. The polymer is carefully fractionated to remove low molecular weight fragments and provide tight mono-modal distribution which results in extremely low bleed.