Puritan® Cotton Tipped Applicator, Puritan Medical Products
Supplier: Puritan Medical Products
860-PPC 860PPCSOLIDHM 860PPCSOLIDHM
CA82001-288CS 1515 CAD
CA82001-288 CA82030-620 CA87006-046 82030-620 87006-046
Puritan® Cotton Tipped Applicator, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent cotton tipped applicator is produced with high quality USP grade cotton which is carded and coiled onsite to maintain consistency
Ideal for cleaning electronics and general-purpose cleaning.
Ordering information: Packaged 100 swabs per bag, 10 bags per pack, 10 packs per case.
Order Now
SPECIFICATIONS
Environmentally Preferable