Microwave Vial Kits, Chemglass
Supplier: Chemglass
CG-4920-01 CG-4920-02 CG-4920-10 CG-4920-04
89041-320EA 1333.6 CAD
89041-320 89041-322 89041-324 10170-366
Microwave Vial Kits, Chemglass
Vials
Vials are manufactured from Type I, Class A borosilicate glass to withstand pressure up to 300 psi.
Ordering information: Seals and stir bars are supplied unassembled.
Packaging: Kit includes 100 vials, 100 aluminum seals with PTFE-faced silicone septa, and 100 PTFE stir vanes.