NextGen™ V Vial® Crimp-Top Sample Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W986212NG W986214NG W986216NG W986217NG W986219NG W986211NG W986213NG
89085-404CS 289.46 CAD
89085-404 89085-408 89085-410 89085-412 89085-414 66021-056 89085-406
NextGen™ V Vial® Crimp-Top Sample Vials, Wheaton, DWK Life Sciences
Vials
Sturdy crimp-top vials have a conical interior bottom for maximum retrieval of content.
Vials can be used for small-scale reactions, centrifugation, storage, and shipping. They are ideal for use in the biochemical, pharmaceutical, and radiochemical industries. Fire-glazed bottom protects against glass particulates. Low-particulate packaging protects against contamination during transportation and storage. Clear vials are made from Wheaton 33® low extractable borosilicate glass to safeguard against a change in product pH caused by alkali, which can be extracted by ordinary glass.
Certifications: Vials conform to ASTM Type I, Class A, and USP Type I requirements.