Rtx®-624 GC Columns, Restek
Supplier: Restek
40925 10968 10969 10972 10973 10970 10974 10971 10975 40924 10971-126 10968-124 10970-600 10971-600 10970-125 10972-125
RK40925EA 2331.21 CAD
RK40925 RK10968 RK10969 RK10972 RK10973 RK10970 RK10974 RK10971 RK10975 10058-088 10059-984 10059-986 10059-988 10059-990 10849-164 10849-652 10849-654 10849-708 10849-796 10850-116 10850-344 10850-360 10850-378
Rtx®-624 GC Columns, Restek
Chromatography Columns
These gas chromatography columns are designed for the analysis of volatile organic compounds using a GC/MS system.
  • Well-suited for EPA method 524.2
  • Application-specific columns for volatile organic pollutants
  • Rtx®-624 phase produces greater than 90% resolution of the first six gases in EPA methods 8260 and 524.2
Equivalent to USP G43 phase
Phase composition: crossbonded 6% cyanopropylphenyl, 94% dimethylpolysiloxane. Temperature range: −20 to 240 °C (−4 to 464 °F). Recommended for EPA methods 502.2, 524.2, 601, 602, 624, 8010, 8020, 8240, and 8260.
More Product Information