Stabilwax®-DA Phase Columns, Restek
Supplier: Restek
11009 11024 11027 11007 11008 11021 11025 11026 11035 11036 11039 11041 11043 11051 11055 11068 11020 11023 11028 11038 11040 11042 11052 11054 11057 11058 11062 11005 11022 11037 11065
RK11009EA 1326.42 CAD
RK11009 RK11024 RK11027 RK11007 RK11008 RK11021 RK11025 RK11026 RK11035 RK11036 RK11039 RK11041 RK11043 RK11051 RK11055 RK11068 RK11020 RK11023 RK11028 RK11038 RK11040 RK11042 RK11052 RK11054 RK11057 RK11058 RK11062 RK11005 RK11022 RK11037 RK11065
Stabilwax®-DA Phase Columns, Restek
Chromatography Columns
Application-specific columns for underivatized and inorganic acids are resistant to oxidative damage.
  • Long-lasting columns
  • Analyze acidic compounds without derivatization
  • Better peak shapes for high molecular weight acids
Columns have an acidic functionality incorporated into the polymer structure, permitting analysis of acidic compounds without derivatization.
Phase composition: crossbonded Carbowax® (100% polyethylene glycol). Temperature range: 40 to 240/250 °C (104 to 464/482 °F).
This significantly reduces adsorption of acids and increases sample capacity for volatile free acids. Columns last longer and provide better peak shapes for high molecular weight acids. Equivalent to USP G25 and G35 phases. For Agilent GCs equipped with split/splitless inlets.