Rtx®-5 Amine Columns, Restek
Supplier: Restek
12353 12354 12352 12355 12366 12369 12382 12385 12338 12351 12350 12323 12335 12354-600 12320
RK12353EA 1961.5 CAD
RK12353 RK12354 RK12352 RK12355 RK12366 RK12369 RK12382 RK12385 RK12338 RK-12351 RK12350 10058-376 10060-032 10803-182 10849-492 10849-600 10849-632 10849-636 10849-764 75849-890
Rtx®-5 Amine Columns, Restek
Chromatography Columns
These base-deactivated, low-polarity columns are designed for the analysis of basic compounds such as alkylamines, diamines, triamines, ethanolamines, and nitrogen-containing heterocycles, as well as neutral compounds, adsorptive compounds with oxygen groups susceptible to hydrogen bonding, and mildly acidic compounds such as phenol.
  • Eliminate the need to derivatize basic compounds
  • Low-polarity column
  • Internal dimensions and film thickness vary between models
Rtx®-5 Amine columns eliminate the need to derivatize the basic compounds before analysisMore Product Information