Rtx®-35 GC Columns, Restek
Supplier: Restek
10440 10420 10424 10437 10438 10439 10453 10455 10470 10485 10421 10423 10435 10454 40410
RK10440EA 1710.96 CAD
RK10440 RK10420 RK10424 RK10437 RK10438 RK10439 RK10453 RK10455 RK10470 RK10485 RK10421 RK10423 RK10435 RK10454 RK40410 10850-698
Rtx®-35 GC Columns, Restek
Chromatography Columns
For analysis of pesticides, PCBs, amines, nitrogen-containing herbicides.
  • Mid-polarity column
  • Different elution order from Rtx®-5
  • Internal dimensions and film thickness vary between models
Other applications include the analysis of sterols, rosin acids, and phthalate esters. A mid-polarity column with a different elution order than the Rtx-5. Phase composition: crossbonded 35% diphenyl, 65% dimethylpolysiloxane. Recommended for EPA method 608.