Synthware Distilling Columns, Vigreux, Kemtech America
Supplier: Kemtech America
C101075 C101100 C101420 C101922 C109130 C102441 C104150 C102442 C102443
60000-764EA 143.69 CAD
60000-764 60000-766 60000-768 60000-770 60000-772 60000-776 60000-778 60000-780 60000-782
Synthware Distilling Columns, Vigreux, Kemtech America
Distillation Components Distillation Columns
Column length is measured from the highest to lowest indent in the column.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Vigreux indentations
Vigreux column features various downward facing indentations that drastically increase surface area.