Automatic Liquid Handling Syringes, H-XL
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2607711 2645017 2645037 2645038 2645058 2607511 2607013 2607033 2607053 2607811 2607911 2645077 2645078 2645918 2607073
77506-238EA 520.94 CAD
77506-238 77506-402 77506-404 77506-406 77506-408 77506-220 77506-158 77506-176 77506-190 77506-252 77506-264 77506-410 77506-412 77506-414 77506-200
Automatic Liquid Handling Syringes, H-XL
Syringes Syringes for Chromatography
XL series syringes are designed to work with most industry standard pump designs e.g. Tecan Cavro.
  • Luer or ¼-28 threaded end caps
  • Broad variety of syringe interfaces
  • Seals available in PTFE, UHMWPE, or custom UHMWPE blends
  • Customized designts and materials to suit application requirements
  • Stroke 60 mm
Glass barrel: Borosilicate 3.3
Front fitting: Glass barrel: Borosilicate 3.3