PYREX® Vacuum Traps, Corning
Supplier: Corning
7729-28 7729-32 7729-38 7729-41 7729-51 7729-48
89091-280CS 2121.24 CAD
89091-280 89091-338 89091-348 89091-382 89091-468 55096-188
PYREX® Vacuum Traps, Corning
Vacuum Traps
These cold traps are made of PYREX® borosilicate glass, clear [ST] joints on both outer and inner joints. The top and side tubulations are the same Ext.Ø. as the centre tube and have a length of 75 mm.