VWR® Tube Racks, PTFE
Supplier: VWR International
89026-358 89026-360 89026-364
89026-358EA 521.59 CAD
89026-358 89026-360 89026-364
VWR® Tube Racks, PTFE
Racks Tube Racks
PTFE racks are useful in corrosive environments where commonly used materials tend to crack or corrode.
  • Tube racks feature a durable PTFE construction
  • Stable support for tubes during chemical reactions
  • Autoclavable
Perfect for supporting tubes during chemical reactions or when storing. Ideal for use between -200°C to 260°C (-328 to 500 °F).