Synthware Vacuum Trap with Serrated Connections, Kemtech America
Supplier: Kemtech America
V163024 V163229 V164034 V164050 V165045 V165050 V165550
60004-718EA 208.55 CAD
60004-718 60004-720 60004-722 60004-724 60004-726 60004-728 60004-730
Synthware Vacuum Trap with Serrated Connections, Kemtech America
Vacuum Traps
With [ST] joints for easy removal of condensate.
  • Heavy wall Borosilicate construction
  • 55/50 Connecting joint
  • 10 mm Center tube
  • Barbed hose connections
Serrated hose connections for the side tube have an O.D. of 13 mm at the smallest serration, and 16 mm at the largest serration. The center tube hose connection is always 10 mm for easy connection to manifold bank tubes of the same diameter.