Synthware Vacuum Trap, Kemtech America
Supplier: Kemtech America
V103029 V103834 V104045 V105045 V105050 V105550
60004-702EA 196.95 CAD
60004-702 60004-704 60004-706 60004-708 60004-710 60004-712
Synthware Vacuum Trap, Kemtech America
Vacuum Traps
Traps feature [ST] joints for easy removal of condensate
  • Heavy wall borosilicate construction
  • 55/50 connecting joint
  • 10 mm center tube
Center tube connection has an O.D. of 10mm. Side tube hose connection is 13mm. Body length is measured below the joint.
Order Now
SPECIFICATIONS
Environmentally Preferable