Mouse MD's™ Meloxicam Bacon Flavor
Supplier: Bio-Serv
Mouse MD's™
MD275-0125 MD275-0125
CA76353-776EA 142.04 CAD
CA76353-776 76353-776
Mouse MD's™ Meloxicam Bacon Flavor
Animal Supplies Animal Feed
Meloxicam is an effective anti-inflammatory drug used in a variety of species including rodents.
Meloxicam MD’s are highly palatable and encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and injurious post-operative injections.
Meloxicam MD’s can be customized to include antibiotics such as Baytril to provide both medications easily and affordably.