Dry Ice Cold Trap, Welch®, Gardner Denver
Supplier: Gardner Denver
1420H-14
55007-176EA 3060.4 CAD
55007-176
Dry Ice Cold Trap, Welch®, Gardner Denver
Vacuum Traps
Dry ice cold trap features a removable 2.8L (3qt.) center well for dry ice/alcohol slurry (do not use acetone) and a visible trapping surface
Trap can maintain cold temperature up to twelve hours depending on vapor load.
ISO size: 12.7 mm (1/2") I.D. Hose