Syringes for Automatic Liquid Handling, XP-XL
Supplier: INNOVATIVE LABOR SYSTEME GMBH
2676015 2676020 2676035 2676815 2676819 2676915 2676916 2676055 2676057 2676075 2676615 2676715
77506-478EA 141.11 CAD
77506-478 77506-480 77506-484 77506-508 77506-510 77506-514 77506-516 77506-490 77506-492 77506-496 77506-500 77506-504
Syringes for Automatic Liquid Handling, XP-XL
Syringes Syringes for Chromatography
The XP-XL syringes for automatic liquid handling.
  • Front fitting: ¹/₄" to 28 UNF
  • Stroke: 30 mm
  • Glass barrel: Borosilicate 3.3
PTFE-seal: Chemically resistant; heavy duty syringes.
HDPE-seal: Long lifetime; good resistance to strong buffers and other aqueous solutions.