Synthware Chromatography Columns, Modified, [ST] 24/40 Bottom Joint, Kemtech America
Supplier: Kemtech America
F234020C F234030C F234030M F236040C F238530C F238540C
60001-982EA 334.82 CAD
60001-982 60001-986 60001-988 60001-998 60002-006 60002-010
Synthware Chromatography Columns, Modified, [ST] 24/40 Bottom Joint, Kemtech America
Chromatography Columns
Short chromatography columns with bottom inner [ST]24/40 joint and hose connection for vacuum.
  • Heavy wall Borosilicate construction
  • Ground glass joints
  • Coarse or medium fritted disc available
Column length is measured from top of column to fritted disc.