Synthware Filter Funnel with Fritted Disc and Vacuum Valve, Kemtech America
Supplier: Kemtech America
F371460M F374200C F374200M
60003-832EA 455.77 CAD
60003-832 60003-834 60003-836
Synthware Filter Funnel with Fritted Disc and Vacuum Valve, Kemtech America
Funnels Filtration Funnels Filter Funnels
Funnels are equipped with a 0 to 4 mm high vacuum valve and a 2 mm glass stopcock that functions as a constant pressure retention device.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Glass stopcock on top, Hi Vac Valve below
  • Coarse or medium fritted disc available
Available with coarse or medium porosity.