Stainless Steel Test Tube Racks, BrandTech
Supplier: BrandTech
B8214 B8222 B8290 B8291 B8293 B8210 B8212 B8292 B8220
80080-298EA 132.29 CAD
80080-298 80080-288 80080-300 80080-294 80080-284 80080-296 80080-290 80080-292 80080-282
Stainless Steel Test Tube Racks, BrandTech
Racks Tube Racks
Polished Type 304 stainless steel racks reduce porosity and provide excellent corrosion resistance in harsh atmospheres.
  • Racks made of 304 stainless steel for long service life
  • Highly polished surfaces for corrosion resistance
  • Z-style racks feature one-piece construction
Bochem stainless test tube racks are made of highly polished 304 (18/10) stainless steel for high corrosion resistance. Standard racks feature a robust welded construction, while the Z-style racks feature one-piece construction for economy and ease of cleaning.