Synthware Filter Funnel with Fritted Disc, Kemtech America
Supplier: Kemtech America
F973014C F973014M F973024C F973024M
60003-818EA 397.28 CAD
60003-818 60003-820 60003-822 60003-824
Synthware Filter Funnel with Fritted Disc, Kemtech America
Funnels Filtration Funnels Filter Funnels
Funnels feature two 2mm glass stopcocks
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Glass stopcock
  • Coarse or medium fritted disc available
Available with coarse or medium porosity.