Inoculating Loops and Needles, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
731101 731161 731165 731170 731175 731180 731181 731185 731171
82051-136CS 326.99 CAD
82051-136 82051-138 82051-140 82051-142 82051-146 82051-148 82051-150 82051-152 82051-144
Inoculating Loops and Needles, Sterile, Greiner Bio-One
Spreaders Inoculating Loops and Needles
Inoculating loops allow easier bacterial collection and inoculation.
  • Color-coded
  • Polystyrene construction
  • Sterile
Inoculating needles are ideal for picking up single colonies.
Order Now
SPECIFICATIONS
Environmentally Preferable