Syringes for Automatic Liquid Handling, TLL-X

Supplier: INNOVATIVE LABOR SYSTEME GMBH
2607717 2607617 2643058 2643077 2643078 2643717 2643817 2643818 2643917 2643017 2643018 2643037 2643038 2643057 2607017 2607037 2607817 2607917 2607057 2607077
77506-244EA 600.51 CAD
77506-244 77506-232 77506-370 77506-372 77506-374 77506-378 77506-380 77506-382 77506-384 77506-360 77506-362 77506-364 77506-366 77506-368 77506-164 77506-182 77506-258 77506-270 77506-194 77506-206
Syringes for Automatic Liquid Handling, TLL-X
Syringes Syringes for Chromatography
The TLL-X series syringes are used for Tecan* Sampler 505/510, RSP 5031/5032/5051/5052/5071/5072/8051, Abbott* TDX/ADX/IMX/Cell Dyn 1300/1600/300/1700, Hamilton* Microlab, Horiba* ABX Pentra 400, ISA Biologie* ISA MAT (HSD), Kratel* Partoscope FD.

  • Front fitting: Luer
  • Plunger top: Button with plunger stop, 6 to 32 UNC
  • Stroke: 60 mm
  • Plunger: PTFE-seal
  • Glass barrel: Borosilicate 3.3
  • Precision: <±1% of the volume

PTFE-seal: Chemically resistant; heavy duty syringes.

PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.

HDPE-seal: Long lifetime; good resistance to strong buffers and other aqueous solutions.

Applications: For sample dosage, diluters and dispensers, dosage of aggressive fluids or luer hub needles and connector.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR