Nunc® Inoculating Loops and Needles, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
251586 254437 253287 254410 253988 254399 254410
CA50806-462CS 2133.27 CAD
CA50806-462 CA50806-465 CA50806-472 CA50806-473 CA50806-481 CA50806-484 50806-473
Nunc® Inoculating Loops and Needles, Sterile, Thermo Scientific
Spreaders Inoculating Loops and Needles
These polystyrene loops and needles are manufactured to very close tolerance levels and surface treated for enhanced droplet adhesion
They provide problem-free streaking with smooth loop surfaces. Ideal for semi-quantitative inoculating procedures. Disposable, nontoxic, and color-coded for product identification. Sterilized by irradiation.
Ordering information: Supplied in easy peel-open bags of 12 or 50.
Order Now
SPECIFICATIONS
Environmentally Preferable