Synthware Reaction Tube Flask with High Vacuum Valve, Kemtech America
Supplier: Kemtech America
F561025 F561050 F561100 F564100 F564250 F564350
60003-280EA 174 CAD
60003-280 60003-282 60003-284 60003-286 60003-288 60003-290
Synthware Reaction Tube Flask with High Vacuum Valve, Kemtech America
Reaction Vessels Reaction Vessels, Flask Shape
Reaction tube flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Teflon® covered high vacuum valve included
Valve size: 0 to 4 mm.