Synthware Reaction Flasks with High Vacuum Valve
Supplier: Kemtech America
F541050 F541950 F541100 F549100 F544100 F541250 F549250 F544250 F544500 F54241L
60003-256EA 244.6 CAD
60003-256 60003-258 60003-260 60003-262 60003-264 60003-266 60003-268 60003-270 60003-272 60003-274
Synthware Reaction Flasks with High Vacuum Valve
Reaction Vessels Reaction Vessels, Flask Shape
Reaction flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Teflon® covered high vacuum valve included
Valve size: 0 to 4 mm.