Syringes for Automatic Liquid Handling, TLL

Supplier: INNOVATIVE LABOR SYSTEME GMBH
2607709 2607809 2607121 2607509 2607609 2641017 2641018 2641037 2641038 2641057 2641058 2641077 2641078 2641618 2641818 2641917 2641918 2607011 2607031 2607051 2607909 2607071 2607091 2607101
77506-236EA 117.08 CAD
77506-236 77506-250 77506-214 77506-218 77506-226 77506-334 77506-336 77506-338 77506-340 77506-344 77506-346 77506-348 77506-350 77506-352 77506-354 77506-356 77506-358 77506-156 77506-174 77506-188 77506-262 77506-198 77506-210 77506-212
Syringes for Automatic Liquid Handling, TLL
Syringes Syringes for Chromatography
These syringes are used for Hamilton* Microlab 400/500/900/M/Digital Dilutor, Behring* BLN Module III, Boehringer* Dilutrend, Cooper/Technicon* RA XT/500/100, Electro* Autoloader I-III, Multistat* III, Mannheim* ES300.

  • Front fitting: Teflon Luer Lock (TLL)
  • Plunger top: Button
  • Stroke: 60 mm
  • Plunger: PTFE-seal
  • Glass barrel: Borosilicate 3.3
  • Precision: <±1% of the volume

PTFE-seal: Chemically resistant; heavy duty syringes.

PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.

HDPE-seal: Long lifetime; good resistance to strong buffers and other aqueous solutions.

Applications: For sample dosage, diluters and dispensers, dosage of aggressive fluids or luer hub needles and connector.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR