FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread

Supplier: AZENTA US, INC
65-7650 65-7651 65-7652 65-7653
77506-896CS 915.33 CAD
77506-896 77506-898 77506-900 77506-902
FluidX 1.6 ml Tri-coded Tubes, 48-format, External Thread
Vials
FluidX 1.6 ml maximum recovery tube is ideally suited for automated workflows using liquid handling, where more sample can be retrieved due to the unique shaped bottom.

  • Sample identification from 1D, 2D and human readable code
  • Capability to work with automation, designed for liquid handlers
  • Suitable for cryogenic storage
  • Made from high-quality virgin PP
  • Available in 48-well format SBS racks or bulk
  • 480 tubes per case or 10 racks per case

Tri-coded tubes offer unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.

Suitable for use with liquid handling automation, whether reformatting, cherry picking, aliquoting an/or normalization. The 1.6 ml maximum recovery tube offers unequaled sample audit traceability, enabling sample tracking and data sharing between multiple users, labs, locations and automation capabilities.

Designed and developed with broad compatibility in mind, the tubes perform without compromise in conjunction with automated barcode reading, capping and sample management systems from Azenta Life Sciences and all other industry-recognized manufacturers.

Storage temperature: Minimum temperature (with screw cap): −196 °C (gas phase liquid nitrogen)

Applications: (For research use only) Liquid handling workflows, biobanking, cryogenic storage, cancer research, cellular and molecular biology, drug discovery, compound libraries, reagent storage, CDMOs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR