Formalin 10%, neutral buffered, VWR®
Supplier: VWR International
|
Warning
|
Synonyms:
Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
77507-018
77507-020
77507-022
77507-024
77507-018CS
229.1
CAD
77507-018
77507-020
77507-022
77507-024
Formalin 10%, neutral buffered, VWR®
Formalin
VWR®, part of Avantor has industry leading Anatomical Pathology reagents available for your lab.
- ISO 13485 Certified
- F-style gallons for easier storage and fewer spills
- Transparency and availability to help you with reliability
Certifications: ISO 13485 Certified.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...