VWR® Large Desiccator Plate
Supplier: VWR
VWR®
J-152
89038-054CS 404.71 CAD
89038-054
VWR® Large Desiccator Plate
Desiccators Desiccators, Plates
Chemical-porcelain plate is glazed on one side and contains eight 30 mm holes.
  • Provided without feet
  • Autoclavable
  • Plate diameter: 230mm